Ward's Formalin-Free Specimens
Economical and Student Safe
Get quality form-free dissection for your classroom at a price that meets your budget with our complete line of formalin-free specimens.
- 100% free of formalin
- Non-sensitizing to skin or eyes
- Comparable in quality and feel to traditional formalin-preserved specimens
- Innovative packing solution further neutralizes residual levels of formaldehyde
- Variety of specimen preparations and packing options