Labconco® Purifier Logic+® Class II, Type A2 Biosafety Cabinets
Through the REDISHIP program, there is only one biosafety cabinet manufacturer that can ship product immediately: Labconco. With 50 years of experience, Labconco is the only name you need when considering biosafety cabinets. The REDISHIP program features Labconco's Logic®+ Class II, Type A2 BSCs which are suitable for all types of biohazards.
Highly featured, the Logic+ combines ergonomics, comfort and safety. A line-of-sight MyLogic™ OS displays cabinet status with clear, concise images and messaging to build user confidence. Constant Airflow Profile™ (CAP) Technology automatically maintains precise airflow, even as the cabinet's HEPA filters load. Users and their samples are kept safe at all times.
The Inclination™ Technology Suite includes a 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and an eye-level mounted color display. A quiet, efficient ECM blower and energy saving LED lights let the cabinet operate efficiently and sustainably for years.
Certifications: NSF/ANSI 49 listed. ETL listed. ADA-compliant (height of controls and receptacles). CE Conformity marking (230 V models). Grade A/ISO 5 conditions per ISO 14644-1 and 2.
See how a Labconco biosafety cabinet can offer you and your samples maximum protection.
MyLogic OS
MyLogic™ OS provides intuitive control of the Logic+ with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. MyLogic™ operating system makes operating your BSC easy.
ECM Blower
Quiet, efficient ECM blower delivers precise volume of air without relying on airflow sensors. Energy saving LED lights reduces cabinet’s carbon footprint. Perfect for labs that want an efficient, sustainable, and maintenance-free cabinet.
Ergonomic Inclination™ Technology Suite
Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display. Designed for maximum user comfort.
Constant Airflow Profile™ (CAP)
Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples.
Never worry if your cabinet is operating at its optimal level. With CAP Labconco BSCs always provide maximum protection.