XPert® Balance Systems & Enclosures
Labconco's™ industry leading XPert Balance Systems and Enclosures provide user protection by keeping powders and particulates contained during manipulations such as weighing. The System features a built-in blower and exhaust HEPA filter and takes up less space on the bench. Vibration isolation allows for steady weighing with a microbalance. Enclosure models are flexible and may be hooked up to a portable exhauster or filtered house exhaust.
Now through the REDISHIP program, Labconco XPert Systems and Enclosures are available within 48 hours.
Certifications: Independently validated by SafeBridge Consultants. Tested to Modified ASHRAE 110. ETL listed. Contact Avantor to see how you can maximize protection from hazardous powders while minimizing space requirements.
XPert® Balance Systems
XPert® Balance Systems provide maximum user protection by keeping powders and particulates contained during weighing operations. Trusted by major global pharmaceutical organizations, XPert Systems keep operators safe from potent active pharmaceutical ingredients (APIs).
XPert® Balance Enclosures
XPert® Enclosures are flexible powder containment hoods that rely on external exhaust or local filtered exhaust systems. Available in sizes up to 8' wide for all types of powder handling processes. Storage cabinet sold separately.
Bag-In/Bag-Out Safe HEPA Filter System
Allows for safe exchange of the HEPA filter without contaminating the surrounding laboratory area.
Built To Last
Tempered safety glass and metal construction offer excellent visibility, ambient light, and long-term durability.