Accurate, Quality Analysis with the Sapphire
The next generation lab imaging platform. This system supports long and short wavelengths of near infrared fluorescence (NIR), red/green/blue (RGB) imaging, chemiluminescent imaging, phosphor imaging as well as optical densitometry (OD) of proteins in stained gels.
Product Features
- Up to four solid state lasers (488, 520, 658 and 784 nm) offering ultimate excitation sensitivity
- Photomultiplier tube (PMT) for fluorescence and phosphor imaging, avalanche photodiodes (APD) for near-infrared imaging and a CCD sensor for chemiluminescent and visible imaging
- Ultra-wide dynamic range for imaging and quantifying low and high abundance samples simultaneously
- Image resolution down to 10 microns for high-quality image analysis
- Full integration with Sapphire Capture and AzureSpot software programs for perfect imaging and accurate analysis every time
Sapphire NIR
Excitation wavelengths at 685 nm and 784 nm
Sapphire RGB
Excitation wavelengths at 488 nm, 520 nm and 658
Sapphire RGBNIR
Excitation wavelengths at 488 nm, 520 nm, 658 nm and 784 nm
Sapphire PI
Excitation wavelength at 658 nm