Advanced Imaging Systems
The Azure 300-600 offer imaging capabilities for chemiluminescence, UV, RGB, NIR, blue light, and white light, depending on the model that fits your needs. The 600 offers the most flexible imaging options, but each model can be upgraded to suit your lab’s needs as they change.
All-in-One Imager for Gels and Blots
Western blot, western blotting, gel doc, gel documentation system, imager
Chemiluminescent Imaging with Color Markers
Overlay a color marker with the chemiluminescent signal for ease of documentation and molecular size determination.
Visible Fluorescence
Chemi and RGB imaging made easy with three-channel RGB excitation for Cy5/Cy3/Cy2 Western applications.
For Two-color Fluorescent Western Blots
Everything you need to perform a single-color fluorescent Western using your own primary antibody and protein sample of your choice.