xTIP Pipette tips for Rainin® LTS® style pipettes
Filtered xTIP Pipette tips
Racked Filtered
xTIP® Pipette Tips are designed for Rainin® LTS-Style Pipets with Biotix® technology. FlexFit Technology lowers the insertion forces required to achieve a secure seal. X-Resin®, and Blade® technologies Ensure clean, accurate delivery every time. xTIP pipette tips are manufactured with a protective polyethylene filter matrix that nearly eliminates aerosol contamination. The tips arrive in the new Biotix rack which uses 35-60% less plastic than competitors. Lift off lid.
Click here to learn more about our LTS Compatible xTIP
Reload Filtered
Coming soon! The filter tip reload is cost effective and engineered with convenience in mind. The reload consists of 2 snap cards of 96 tips each and easily clicks into your Biotix rack directly from the packaging.
Non-Filter xTIP Pipette Tips
Racked Non-Filtered
xTIP® Pipette Tips are designed for Rainin® LTS-Style Pipets with Biotix® technology. FlexFit Technology lowers the insertion forces required to achieve a secure seal. X-Resin®, and Blade® technologies Ensure clean, accurate delivery every time. xTIP pipette tips are tested and certified free of RNase, DNase, Pyrogens, and PCR inhibitors. The tips arrive in the new Biotix rack which uses 35-60% less plastic than competitors. Lift off lid.
Click here to learn more about our LTS Compatible xTIP
CleanPak Reload Non-Filtered
CleanPak is the reload solution for the lab that is looking to maximize lab space, reduce their environmental footprint and has a tight budget to work with. CleanPak is packaged in a hermetically sealed bag and comes in both sterile and non-sterile options. The user has the option to reload one snapcard at a time and reseal for later use or rack all 10 snapcards at once.
Click to browse our CleanPak reload options
TipEject Reload - Non-Filter
The TipEject allows you to reload your racks without coming in contact with the tips. It provides a clean reload solution with easy one-handed error free loading. TipEject’s rigid plastic covering shields tips from the laboratory environment from the top down, keeping contamination out. Simply place the TipEject sleeve over the rack and press down to release your tips into the rack base without touching the snap card or the tips.
Click here for more information on the TipEject reload