Cobra® Pipette
DESIGNED FOR COMFORT AND ACCURACY.
The Cobra Manual Pipettor System combines air displacement technology with the latest ergonomic research for the most comfortable pipetting imaginable. The low pipetting forces required reduce the risk of repetitive stress disorder, and the built-in control settings ensure accuracy in ranges from 0.2 μL to 1000 μL. The Cobra pipette is available in single and multichannel models.
ERGONOMICS
- Easily used by left-hand or right-hand users thanks to the adjustable tip-ejector button
- Extra-long finger hook rests securely into hands of all sizes for a firm grip
CONSTRUCTION
- Light-weight design falls within the ideal weight range
- Fully autoclavable at 121° C, 1 atm for 20 minutes
- Stainless-steel piston provides precision and durability
- Full 12-month warranty
Cobra® Pipette