Human Recombinant RH IL-7 (from E. coli)
Supplier: VWR International
77753-374 77753-376 77753-378
77753-374EA 2035.15 USD
77753-374 77753-376 77753-378
Human Recombinant RH IL-7 (from E. coli)
Proteins and Peptides
IL-7 promotes the stimulation of multipotent (pluripotent) hematopoietic stem cells into lymphoid progenitor cells. IL-7, along with IL-2, 15, and 21, is a member of the common gamma (γc) chain cytokine family.
Secreted by bone marrow and thymic stromal cells, dendritic cells, intestinal epithelial cells, hepatocytes, and keratinocytes
Signals through its receptor (IL-7R) to promote the differentiation of hematopoietic stem cells into T cells, B cells, and natural killer cells
Is a regulator of intestinal mucosal lymphocyte proliferation
Promotes favorable phenotypes in adoptive cellular therapies such as CAR-T cell therapy when combined with IL-15. The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
  • High quality
  • Low endotoxin
  • Lot-to-lot consistency
  • Animal free
  • Manufactured following GMP guidelines
Host cell protein: ≤30 ng/ml, Host cell DNA: ≤0.01 ng/ml, Sterility testing: No growth, Mycoplasma: Negative.
Caution:
For research use or further manufacturing only.