Human Recombinant PLGF-3 (from E. coli)
Supplier: VWR International
77750-964 77750-966 77750-968 77750-970 77750-972
77750-964EA 855.98 USD
77750-964 77750-966 77750-968 77750-970 77750-972
Human Recombinant PLGF-3 (from E. coli)
Proteins and Peptides
Placenta growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family and promotes endothelial cell growth and angiogenesis. PlGF is secreted as a homodimer and may also form a heterodimer with VEGF. PlGF is detected in the placenta, heart, lungs, thyroid, and adipose tissues. Circulating PlGF levels are correlated with colorectal and renal cancers, as well as atherosclerosis and ischemic heart disease. There are four alternatively spliced PlGF isoforms (PlGF-1, PlGF-2, PlGF-3, and PlGF-4), each with unique secretion patterns and heparin-binding affinities. PlGF-3 lacks a heparin-binding domain and signals through the VEGFR-1 receptor.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.