Human Recombinant M-CSF (from E. coli)
Supplier: VWR International
77750-054 77750-056 77750-058 77750-060
77750-054EA 855.98 USD
77750-054 77750-056 77750-058 77750-060
Human Recombinant M-CSF (from E. coli)
Proteins and Peptides
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types. M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy. Human M-CSF shows activity on mouse cells; however, mouse M-CSF shows no activity on human cells.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.