L-Methionylglycine

Supplier: ALADDIN SCIENTIFIC

M121443-5G M121443-100MG M121443-1G M121443-500MG
ALM121443-5GEA 855.9 USD
ALM121443-5G ALM121443100MG ALM121443-1G ALM121443500MG
L-Methionylglycine
Proteins and Peptides

  • EINECS: 238-487-9
  • Melting Point: 212 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR