Albumin human
Supplier: ALADDIN SCIENTIFIC
H304436-1G H304436-2.5G H304436-500MG H304436-5G
ALH304436-1GEA 131.9 USD
ALH304436-1G ALH304436-2.5G ALH304436500MG ALH304436-5G
Albumin human
Proteins and Peptides
Product Introductionrecombinant, expressed in rice, suitable for cell culture, low endotoxin;It is a kind of recombinant human albumin (RHA) without animal origin. It is designed for cell culture applications and is a powerful medium supplement with superior performance to human albumin, bovine serum albumin and recombinant human albumin from other sources.It brings stable and consistent performance rather than variables related to hydrolysates and extracts. It is an ideal medium for Chinese hamster ovary (CHO), hybridoma, Vero, stem cell and primary cell culture medium.The recombinant variant is recommended for absorption, transport, metabolism and excretion (ADME) pharmacology; cell culture; drug delivery; and cell cryopreservation.