L-Methionylglycine

Supplier: TCI
M0250-1G
TCM0250-001GEA 235.62 USD
TCM0250-001G
L-Methionylglycine
Proteins and Peptides
Other Product Names / Synonyms: H-Met-Gly-OH

  • Melting point: 212 °C
  • Purity (Nonaqueous titration): Min. 95.0%
  • Specific rotation [a]20/D: 85.0 to 90.0° (C=2, Water)
  • Solubility in water: almost transparency
  • EINECS: 238-487-9
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR