L-Alanyl-L-Glutamine

Supplier: TCI
A1261-1G
TCA1261-001GEA 24.85 USD
TCA1261-001G
L-Alanyl-L-Glutamine
Proteins and Peptides
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Ala-Gln-OH

  • Specific rotation [a]20/D: 10° (C=5, H₂O)
  • Melting point: 215 °C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR