Absorber Set
Supplier: SPECTRUM TECHNIQUES, LLC
RAS-20
470123-270KT 481 USD
470123-270
Absorber Set
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Perfectly Suited for Use with Our Nuclear Labs.
  • Complete set for radiation studies
  • Includes storage case
This set of 20 calibrated radiation absorbers covers the range from 4.5 to 7400 mg/cm2 and consists of aluminum, plastic and lead plates. The absorber are 2.75 in2 and are designed to fit all Spectrum Techniques detector stand assemblies.
The set is suitable for absorption studies with Alpha, Beta and Gamma radiation.
Delivery information: This item is Item only available to US Customers.
Packaging: One set contains twenty items.