VWR® Glycylglycine (Gly-Gly)

Supplier: Avantor
0137-1KG 0137-100G 0137-250G
97061-066CS 1097.21 USD
97061-066 97061-068 97061-070
VWR® Glycylglycine (Gly-Gly)
Proteins and Peptides
Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH.

  • Loss on drying: ≤0.2%
  • Solubility (10%, water): Pass
  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning

Glycylglycine (Gly-Gly) contains ≤0.02% ammonia, ≤0.0001% arsenic, ≤0.2% ash, ≤0.02% chloride, ≤0.002% heavy metals, ≤0.001% iron and ≤0.03% sulfate.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR