Albiglutide (Acetate)
Supplier: ALADDIN SCIENTIFIC
A275359-25MG A275359-2MG A275359-5MG
ALA275359-25MGEA 1407.81 USD
ALA275359-25MG ALA275359-2MG ALA275359-5MG
Albiglutide (Acetate)
Proteins and Peptides
Albiglutide is a recombinant fusion protein consisting of two copies of a 30-amino-acid sequence of modified human GLP-1 (fragment 7 - 36), modified with a glycine substituted for the naturally occurring alanine at position 8 in order to augment resistance to DPP-4.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf30c78c6aa6cfaaeac608862544f352f9ad862b&vid=cf30c78c6aa6cfaaeac608862544f352f9ad862b&prid=web&clid=SAPDAM