Human ACTH (18-39) (TFA)
Supplier: ALADDIN SCIENTIFIC
A276099-1MG A276099-5MG
ALA276099-1MGEA 94.91 USD
ALA276099-1MG ALA276099-5MG
Human ACTH (18-39) (TFA)
Proteins and Peptides
Adrenocorticotropic Hormone (ACTH) (18-39), human TFA is a corticotrophin-like peptide produced in melanocytotropic cells in the middle lobe of the pituitary glandAdrenocorticotropic Hormone Fragment 18 - 39 human (CLIP) is a hormone which stimulates the release of insulin. Studies indicate that fragments of ACTH can antagonize opiate receptors. Adrenocorticotropic Hormone Fragment 18 - 39 human is a tool used to monitor rapid eye movement sleep in rats due to its endogenous hypnogenic factors. Furthermore, CLIP can be used as an intermediate in the production of ACTH22-39.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d7218df60866077d3fd9b8a05f4b69222f965ff&vid=5d7218df60866077d3fd9b8a05f4b69222f965ff&prid=web&clid=SAPDAM