Ala-Pro Hydrate

Supplier: ALADDIN SCIENTIFIC

A167052-1G A167052-250MG A167052-5G
ALA167052-1GEA 54.06 USD
ALA167052-1G ALA167052250MG ALA167052-5G
Ala-Pro Hydrate
Proteins and Peptides

  • GHS Signal word: Warning
  • Melting point: 171 °C

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03f0d78d3bfef5a3c0ae9c36da817c2f648974df&vid=03f0d78d3bfef5a3c0ae9c36da817c2f648974df&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR