Soybean Trypsin Inhibitor, ACF
Supplier: STEMCELL Technologies
07457 07458
MSPP-07457EA 567.08 USD
MSPP-07457 MSPP-07458
Soybean Trypsin Inhibitor, ACF
Cell Culture Supplements and Additives
For inhibition of trypsin activity.
  • Keep cultures free of animal-derived materials with a soybean-sourced trypsin inhibitor
  • Effectively inhibits trypsin, as well as human and bovine thrombin, plasmin, and leukocytic proteases
Inactivate trypsin with Soybean Trypsin Inhibitor, Animal Component-Free (ACF). This trypsin inhibitor consists of a single polypeptide chain with two disulfide bridges. It is able to inhibit chymotrypsin to a lesser extent, but is ineffective against esterolytic, proteolytic, or elastolytic activities of porcine elastase (Vered et al.). Soybean Trypsin inhibitor, ACF has also been shown to inhibit human and bovine thrombin, plasmin, and leukocytic proteases.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').