EasySep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kits
Supplier: STEMCELL Technologies
17878 17878RF
MSPP-17878EA 1129.23 USD
MSPP-17878 MSPP-17878RF
EasySep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kits
Cell Separation Media
Column-free immunomagnetic positive selection of human CD14+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kit is designed to isolate CD14+ cells from freshly prepared buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD14 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cellsMore Product Information