EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kits
Supplier: STEMCELL Technologies
17881 17881RF
MSPP-17881EA 1240 USD
MSPP-17881 MSPP-17881RF
EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kits
Cell Separation Media
Immunomagnetic positive selection of human CD15+ cells (e.g. myeloid cells) from whole blood.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit is designed to isolate highly purified CD15+ cells from fresh whole blood by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD15 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube whileMore Product Information