EasySep™ Human CD8+ T Cell Isolation Kits
Supplier: STEMCELL Technologies
17953 100-0710 17953RF
MSPP-17953EA 1466.15 USD
MSPP-17953 MSPP-100-0710 MSPP-17953RF
EasySep™ Human CD8+ T Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human CD8+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 91% purity with high recovery
  • Untouched, viable cells
The EasySep™ Human CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This product can be used in place of the EasySep™ Human CD8+ T Cell Enrichment Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').