EasySep™ Human Basophil Isolation Kits
Supplier: STEMCELL Technologies
17969 17969RF
MSPP-17969EA 1381.54 USD
MSPP-17969 MSPP-17969RF
EasySep™ Human Basophil Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human basophils.
  • Fast, easy-to-use and column-free
  • Up to 98% purity
  • Isolated cells are untouched
The EasySep™ Human Basophil Isolation Kit is designed to isolate basophils from a polymorphonuclear cell-rich fraction by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-basophils and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
The isolation kit is compatible with cells prepared using HetaSep™ sedimentation of fresh peripheral blood. This product replaces the EasySep™ Human Basophil Enrichment Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').