EasySep™ Rat IgM Depletion Kits
Supplier: Stemcell Technologies
18644 18644RF
MSPP-18644EA 1521.54 USD
MSPP-18644 MSPP-18644RF
EasySep™ Rat IgM Depletion Kits
Cell Separation Media
Immunomagnetic depletion of rat IgM+ cells.
  • Fast, easy-to-use and column-free
  • Up to 90% purity (≤10% IgM+)
  • Isolated cells are untouched
The EasySep™ Rat IgM Depletion Kit is designed to deplete IgM+ cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by positive selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').