EasySep™ Human CD15 Positive Selection Kits
Supplier: STEMCELL Technologies
18651 18651RF
MSPP-18651EA 1336.92 USD
MSPP-18651 MSPP-18651RF
EasySep™ Human CD15 Positive Selection Kits
Cell Separation Media
Immunomagnetic positive selection of human CD15+ cells (e.g. Myeloid cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human CD15 Positive Selection Kit is designed to isolate CD15+ cells from ammonium chloride lysed blood or other nucleated cell suspensions by positive selection. Desired cells are targeted with antibody complexes recognizing CD15 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD15 antigen is expressed on monocytes and granulocytes.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').