EasySep™ HLA B Cell Enrichment Kits
Supplier: STEMCELL Technologies
19054HLA 19054HLARF
MSPP-19054HLAEA 1472.31 USD
MSPP-19054HLA MSPP-1954HLARF
EasySep™ HLA B Cell Enrichment Kits
Cell Separation Media
Column-free immunomagnetic isolation of untouched human B cells for HLA analysis.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Isolated cells are untouched
  • Compatible with downstream HLA assays
The EasySep™ HLA B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet and without the use of columns. Desired cells are poured off into a new tube and are ready for serology or flow cytometry crossmatch assays.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').