EasySep™ Human Monocyte Enrichment Kits
Supplier: Stemcell Technologies
19059 19059RF
MSPP-19059EA 1446.15 USD
MSPP-19059 MSPP-19059RF
EasySep™ Human Monocyte Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched human monocytes.
  • Fast, easy-to-use and column-free
  • Up to 95% purity
  • Untouched, viable cells
The EasySep™ Human Monocyte Enrichment Kit is designed to isolate monocytes from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-monocyte cells and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to prevent nonspecific binding of monocytes. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For applications in which CD16+ cells are not removed, we recommend the EasySep™ Human Monocyte Enrichment Kit without CD16 Depletion which isolates cells in as little as 12.5 minutes.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').