EasySep™ Human Myeloid DC Enrichment Kits
Supplier: Stemcell Technologies
19061 19061RF
MSPP-19061EA 1536.92 USD
MSPP-19061 MSPP-19061RF
EasySep™ Human Myeloid DC Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched human myeloid dendritic cells (mDCs).
  • Fast, easy-to-use and column-free
  • Up to 90% purity
  • Untouched, viable cells
The EasySep™ Human Myeloid DC Enrichment Kit is designed to isolate myeloid dendritic cells (mDC) from fresh peripheral blood mononuclear cells or ammonium chloride-lysed leukapheresis by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-mDCs and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').