EasySep™ Human Pan-DC Pre-Enrichment Kits
Supplier: STEMCELL Technologies
19251 19251RF
MSPP-19251EA 1243.08 USD
MSPP-19251 MSPP-19251RF
EasySep™ Human Pan-DC Pre-Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched human pan dendritic cells (DC)
  • Fast, easy-to-use and column-free
  • Up to 80% purity
  • Untouched, viable cells
The EasySep™ Human Pan-DC Pre-Enrichment Kit is designed to pre-enrich all dendritic cells (DCs) (including myeloid and plasmacytoid DCs) from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-pan DCs and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').