EasySep™ Human Naïve CD8+ T Cell Isolation Kits
Supplier: STEMCELL Technologies
19258 19258RF
MSPP-19258EA 1592.31 USD
MSPP-19258 MSPP-19258RF
EasySep™ Human Naïve CD8+ T Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human naïve CD8+ T cells
  • Fast, easy-to-use and column-free
  • Up to 97% purity
  • Untouched, viable cells
The EasySep™ Human Naïve CD8+ T Cell Isolation Kit is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD4, CD14, CD16, CD19, CD20, CD36, CD45RO, CD56, CD57, CD94, CD123, CD244, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
For even faster cell isolations, we recommend the EasySep™ Human Naïve CD8+ T Cell Isolation Kit II which isolates cells in as little as 11 minutes.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').