EasySep™ Human Pan-Granulocyte Isolation Kits
Supplier: STEMCELL Technologies
19259 19259RF
MSPP-19259EA 1510.77 USD
MSPP-19259 MSPP-19259RF
EasySep™ Human Pan-Granulocyte Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human pan-granulocytes
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
The EasySep™ Human Pan-Granulocyte Enrichment Kit is designed to isolate granulocytes (eosinophils, basophils and neutrophils) from normal peripheral whole blood by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-granulocytes and magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells areMore Product Information