EasySep™ Rat T Cell Isolation Kits
Supplier: STEMCELL Technologies
19641 19641RF
MSPP-19641EA 1090.77 USD
MSPP-19641 MSPP-19641RF
EasySep™ Rat T Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched rat T cells
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Rat T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desiredMore Product Information