EasySep™ Direct HLA Crossmatch T Cell Isolation Kits
Supplier: STEMCELL Technologies
19671 19671RF
MSPP-19671EA 872.31 USD
MSPP-19671 MSPP-19671RF
EasySep™ Direct HLA Crossmatch T Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human T cells for HLA analysis
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 99.9% purity of isolated cells
  • Fast, easy-to-use, and column-free
  • Isolated cells are untouched
The EasySep™ Direct HLA Crossmatch T Cell Isolation Kit can be used to isolate highly purified T cells directly from human whole blood, spleen or lymph nodes by immunomagnetic negative selection. This kit targets non-T cellsMore Product Information