EasySep™ Mouse Neutrophil Enrichment Kits
Supplier: STEMCELL Technologies
19762 19762RF
MSPP-19762EA 1213.85 USD
MSPP-19762 MSPP-19762RF
EasySep™ Mouse Neutrophil Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched mouse neutrophils
  • Fast, easy-to-use and column-free
  • Up to 93.7% (blood) and 88.7% (bone marrow) purity
  • Isolated cells are untouched
The EasySep™ Mouse Neutrophil Enrichment Kit is designed to isolate neutrophils from single-cell suspensions of bone marrow or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-neutrophils. Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran. These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').