EasySep™ Mouse CD4+ T Cell Isolation Kits
Supplier: STEMCELL Technologies
19852 19852RF
MSPP-19852EA 1216.92 USD
MSPP-19852 MSPP-19852RF
EasySep™ Mouse CD4+ T Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched mouse CD4+ T cells
  • Fast and easy-to-use
  • Up to 96% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-CD4+ T cells and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
This product replaces the EasySep™ Mouse CD4+ T Cell Enrichment Kit (Catalog #19752) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').