EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kits
Supplier: STEMCELL Technologies
19860 19860RF
MSPP-19860EA 721.54 USD
MSPP-19860 MSPP-19860RF
EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched mouse cells
  • Fast and easy-to-use
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit is designed for the depletion of single or multiple unwanted cell types labeled with biotinylated antibodies. Labeled cells from mouse splenocytes or other tissues are targeted for removal by Streptavidin RapidSpheres™ and separated without the use of columns using an EasySep™ magnet. Desired cells are untouched and poured off into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').