RosetteSep™ Human Mesenchymal Stem Cell Enrichment Cocktails
Supplier: Stemcell Technologies
15168 15128
MSPP-15168EA 1481.54 USD
MSPP-15168 MSPP-15128
RosetteSep™ Human Mesenchymal Stem Cell Enrichment Cocktails
Cell Separation Media
Immunodensity isolation of untouched mesenchymal stem cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
The RosetteSep™ Human Mesenchymal Stem Cell Enrichment Cocktail is designed to isolate mesenchymal stem cells from fresh bone marrow by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3, CD14, CD19, CD38, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The purified mesenchymal stem cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).