RosetteSep™ Human Progenitor Cell Basic Pre-Enrichment Cocktails
Supplier: STEMCELL Technologies
15226 15266
MSPP-15226EA 1469.23 USD
MSPP-15226 MSPP-15266
RosetteSep™ Human Progenitor Cell Basic Pre-Enrichment Cocktails
Cell Separation Media
Immunodensity pre-enrichment of untouched progenitor cells from cord blood.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human Progenitor Cell Basic Pre-Enrichment Kit is designed to pre-enrich hematopoietic progenitor cells from cord blood by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD3, CD11b, CD19 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The hematopoietic progenitor cells are present as an enriched population at the interface between the plasma and the buoyant density medium.
Cells of interest can then be positively selected from the pre-enriched cell population using EasySep™ (e.g. EasySep™ Human CD34 Positive Selection).
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).