RosetteSep™ HLA Total Lymphocyte Enrichment Cocktails
Supplier: STEMCELL Technologies
15283HLA 15263HLA
MSPP-15283HLAEA 4898.46 USD
MSPP-15283HLA MSPP-15263HLA
RosetteSep™ HLA Total Lymphocyte Enrichment Cocktails
Cell Separation Media
Immunodensity isolation of untouched lymphocytes for HLA analysis.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Isolated cells are untouched
The RosetteSep™ HLA Total Lymphocyte Enrichment Cocktail is designed to enrich lymphocytes from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD16, CD36, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (STMC15705) or Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The purified lymphocytes are present as a highly enriched population at the interface between the plasma and the buoyant density medium. The desired cells are immediately ready for flow cytometry crossmatch assays or other downstream applications.
Please Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled 'For Research Use Only'. Please note there have been no changes to the form, function and quality assurances associated with these products.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).