RosetteSep™ Human CD4 Depletion Cocktails
Supplier: Stemcell Technologies
15622 15662
MSPP-15622EA 213.85 USD
MSPP-15622 MSPP-15662
RosetteSep™ Human CD4 Depletion Cocktails
Cell Separation Media
Immunodensity depletion of CD4+ cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human CD4 Depletion Cocktail is designed to deplete CD4+ cells from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD4 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The CD4+ depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).