EasySep™ Human Naïve CD4+ T Cell Isolation Kit II
Supplier: Stemcell Technologies
17555 17555RF
MSPP-17555EA 1466.15 USD
MSPP-17555 MSPP-17555RF
EasySep™ Human Naïve CD4+ T Cell Isolation Kit II
Cell Separation Media
Immunomagnetic isolation of untouched human naïve CD4+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 98% purity
  • Untouched, viable cells
The EasySep™ Human Naïve CD4+ T Cell Isolation Kit II is designed to isolate naïve CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This kit replaces the EasySep™ Human Naïve CD4+ T Cell Enrichment Kit (STMC19155) and EasySep™ Human Naïve CD4+ T Cell Isolation Kit (STMC19555) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).