EasySep™ Human 'Do-It-Yourself' Positive Selection Kit II
Supplier: Stemcell Technologies
17699 17699RF
MSPP-17699EA 1130.77 USD
MSPP-17699 MSPP-17699RF
EasySep™ Human 'Do-It-Yourself' Positive Selection Kit II
Cell Separation Media
Immunomagnetic positive selection of human cells with your own mouse IgG1 antibody.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Human 'Do-It-Yourself' Positive Selection Kit II is designed to select any human cell type of interest with your own mouse IgG1 monoclonal antibody. Cells that express the surface antigen that is recognized by the mouse IgG1 monoclonal antibody are targeted for magnetic separation using EasySep™ Dextran RapidSpheres™ and the EasySep™ Magnet. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding.
This product replaces the EasySep™ Human 'Do-It-Yourself' Selection Kit (STMC18099).
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).