EasySep™ HLA Chimerism Whole Blood Lymphoid Positive Selection Kits
Supplier: Stemcell Technologies
17873 17873RF
MSPP-17873EA 1090.77 USD
MSPP-17873 MSPP-17873RF
EasySep™ HLA Chimerism Whole Blood Lymphoid Positive Selection Kits
Cell Separation Media
Column-free immunomagnetic positive selection of human CD3+ and CD19+ cells (e.g. lymphocytes) cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99.9% purity
  • No columns required
The EasySep™ Human Whole Blood Lymphoid Positive Selection Kit is designed to isolate CD3+ and CD19+ cells from whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD3, CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD3 antigen is expressed on T lymphocytes and NKT cells while the CD19 antigen is expressed on B cells and follicular dendritic cells.
This product replaces the EasySep™ Human Whole Blood Lymphoid Positive Selection Kit (STMC18684) and EasySep™ HLA Whole Blood Lymphoid Positive Selection Kit (STMC18684HLA) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).