Human MET (from HEK293 cells), PE (Phycoerythrin)
Supplier: Sino Biological
10692H86HSP100 10692H86HSP25
77582-866EA 1027.94 USD
77582-866 77582-868
Human MET (from HEK293 cells), PE (Phycoerythrin)
Proteins and Peptides
A DNA sequence encoding the Human MET (NP_000236.2) (Met1-Thr932) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with PE (Excitation = 495 to 566 nm, Emission Max. = 575 nm).