Human EGFR (from HEK293 cells), Alexa Fluor® 488
Supplier: Sino Biological
10001H86HSD100 10001H86HSD25
77581-366EA 1151.54 USD
77581-366 77581-368
Human EGFR (from HEK293 cells), Alexa Fluor® 488
Proteins and Peptides
A DNA sequence encoding the Human EGFR (NP_005219.2) (Met1-Ser645) was expressed with a polyhistidine tag at the C-terminus. The protein is site-specifically labeled with AF 488 (Excitation Max. = 495 nm, Emission Max. = 519 nm).