Water nuclease-free

Supplier: STEMCELL Technologies

Synonyms: Dihydrogen oxide

79001 79002 79002.1
MSPP-79001EA 95 USD
MSPP-79001 MSPP-79002 MSPP-790021006
Water nuclease-free
Water

DNase- and RNase-free water for molecular biology applications.


Nuclease-Free Water is used in various molecular biology applications requiring nuclease-free conditions, such as in processing DNA or RNA (i.e. PCR, cDNA synthesis, or qPCR). This product is sterile-filtered (0.2 µm), free of RNase and DNase activity, endotoxin-free, and not DEPC-treated.


Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Formula: H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR