7-Aminoactinomycin D (7-AAD) cell viability stain

Supplier: STEMCELL Technologies

Synonyms: NSC-2397597-AADActinomy2,7-diamino-4,6-dimethyl-3-oxo-1-N,9-N-bis[7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]phenoxazine-1,9-dicarboxamide

75001 75001.1
MSPP-750011006EA 65.77 USD
MSPP-750011006 MSPP-75001
7-Aminoactinomycin D (7-AAD) cell viability stain
7-Aminoactinomycin D (7-AAD)

7‐AAD (7‐Aminoactinomycin D) is a fluorescent cell viability dye which is excluded from live cells with intact membranes but penetrates dead or damaged cells and binds to double‐stranded DNA with high affinity by intercalating between GC base pairs.


  • Cell viability dye (DNA-labeling dye)


It is often used as an alternative to propidium iodide (PI; Catalog #75002) to differentiate and exclude non-viable cells in flow cytometric analyses, and can be excited using 488 nm (blue), 532 nm (green), and 561 nm (yellow-green) laser lines. Compared to PI, 7-AAD offers the advantage of having minimal overlap in its emission spectra with mostly blue and green, and many red fluorophores, including the commonly used phycoerythrin (PE) and fluorescein isothiocyanate (FITC). 7‐AAD is also used in DNA fluorescence imaging applications to discriminate early and late stages of apoptosis, to study cell‐mediated cytotoxicity, and for chromosome banding analysis. 7‐AAD has been found to exhibit antibacterial properties and exhibits growth‐inhibitory activity against certain types of leukemia and sarcoma.


Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Formula: C₆₂H₈₇N₁₃O₁₆
MW: 1270.43 g/mol
MDL Number: MFCD00077295
CAS Number: 7240-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR