Anti-CD235ab Mouse Monoclonal Antibody [clone: HIR2 (GA-R2)] (PE)

Supplier: STEMCELL Technologies
60111PE.1 60111PE
MSPP-60111PEEA 567.08 USD
MSPP-60111PE MSPP-60111PE1
Anti-CD235ab Mouse Monoclonal Antibody [clone: HIR2 (GA-R2)] (PE)
Antibodies
Mouse monoclonal IgG2b antibody against human CD235ab (glycophorin A/B), PE-conjugated.

  • Red blood cells

The HIR2 (GA-R2) antibody recognizes an epitope common to the N-terminal region of human CD235a (glycophorin A) and CD235b (glycophorin B), homologous type I sialoglycoproteins of the erythrocyte (red blood cell) membrane that bear the antigenic determinants for the MN and Ss blood groups. CD235ab is expressed on early- and late-stage erythroblasts, mature erythrocytes, and erythroid cell lines such as K562 and HEL, but not on other cell types. CD235a is abundantly expressed whereas CD235b is a relatively minor membrane component. The proteins are believed to provide a large mucin-like surface to erythrocytes that acts to minimize aggregation in the circulation. The HIR2 antibody binds with higher affinity to CD235a than CD235b, and agglutinates untreated erythrocytes.

Type:
Antigen: CD235ab
Clonality: Monoclonal
Clone: HIR2 (GA-R2)
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR