Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PerCP-Cy5.5)
Supplier: STEMCELL Technologies
60068PS 60068PS.1
MSPP-60068PSEA 659.15 USD
MSPP-60068PS MSPP-60068PS1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PerCP-Cy5.5)
Antibodies
Rat monoclonal IgG2b antibody against human, mouse, rhesus CD44 (tissue non-specific alkaline phosphatase), PerCP-Cy5.5-conjugated.
  • Mammary cells
  • T cells
The IM7 antibody reacts with CD44 (Ly-24), an ~80 - 95 kDa type 1 transmembrane glycoprotein involved in cell-cell and cell-matrix interactions. CD44 is expressed on the surface of many cells, including leukocytes and hepatocytes, as well as endothelial, epithelial, and mesenchymal cells. Expression levels increase upon activation of T and B cells, and memory cells exhibit a CD44high phenotype. CD44More Product Information